Learn More
Sigma Aldrich Fine Chemicals Biosciences scoves Modfed Dulbeccos500ML
Supplier:  Sigma Aldrich Fine Chemicals Biosciences FG0465500ML
Encompass
This IMDM differs from the original formula because it is supplemented with L-alanyl-L-glutamine dipeptide. Glutamine is notoriously unstable in cell culture. This dipeptide provides a more stable form of glutamine for use in cell cultures.
Catalog No. 50-290-1489