Learn More
SCP Science Tubing, PVC Blue/Blue (15)
Catalog No. NC1760674
Encompass
This item has a minimum qty of 3 per supplier requirements.
PVC Pump Tubing ID: 1.65mmInter-stop distance: 150mm
Catalog No. NC1760674 Supplier SCP Science Supplier No. 022030013
This item has a minimum qty of 3 per supplier requirements.
PVC Pump Tubing ID: 1.65mmInter-stop distance: 150mm