Learn More
Xcessbio SCR7 10MM DMSO (0.598ML)
Supplier:  Xcessbio M600822S
Encompass_Preferred
Product Information Molecular Weight: 334.39 Formula: C18H14N4OS Purity: >= 98%. Solubility: DMSO up to 50 mM Chemical Name: 5,6-bis((E)-benzylideneamino)-2-mercaptopyrimidin-4-ol Storage: Powder: 4oC 1 year. DMSO: 4oC 3 month; -20oC 1 year. Biological activity: SCR7 is a potent and selective inhibitor of non-homologous end joining (NHEJ). It inhibits joining of DSBs in cell-free DNa repair system, blocks Ligase IV-mediated joining by interfering with its DNa binding but not that of T4 DNa Ligase or Ligase I, thereby leading to accumulation of DSBs within the cells, culminating into cytotoxicity. SCR7 inhibits NHEJ in a Ligase IV-dependent manner within cells, and activates the intrinsic apoptotic pathway. More importantly, SCR7 impedes tumor progression in mouse models, and when co-administered with DSB-inducing therapeutic modalities it enhances their sensitivity significantly. In addition, SCR7 can promote the efficiency of HDR 4-5-fold for CRISPR editing in both h 1533426-72-0
Catalog No. NC0769142