Learn More
Selleck Chemical LLC SCR7 5mg 14892-97-8
SDP
Supplier:  Selleck Chemical LLC S77425MG
Encompass_Preferred
SCR7 is a specific DNA Ligase IV inhibitor, which blocks nonhomologous end-joining (NHEJ). It increases the efficiency of HDR-mediated genome editing up to 19-fold using CRISPR/Cas9 in mammalian cells and mouse embryos. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0973358