Learn More
TARGETMOL CHEMICALS INC SCR7 PYRAZINE 10MG
SDP
Catalog No. 502577313
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SCR7 pyrazine (SCR7) enhances CRISPR-Cas9-mediated homology-directed repair (HDR) efficiency in vitro up to 19-fold. Inhibits nonhomologous end-joining (NHEJ). purity: 99%
Catalog No. 50-257-7313 Supplier TARGETMOL CHEMICALS INC Supplier No. T172410MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SCR7 pyrazine (SCR7) enhances CRISPR-Cas9-mediated homology-directed repair (HDR) efficiency in vitro up to 19-fold. Inhibits nonhomologous end-joining (NHEJ). purity: 99%