Learn More
Medchemexpress LLC HY-107845 5mg Medchemexpress, SCR7 pyrazine CAS:14892-97-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1078455MG
Encompass_Preferred
Medchemexpress, HY-107845 5mg SCR7 pyrazine CAS:14892-97-8 SCR7 pyrazine is a DNA ligase IV inhibitor that blocks nonhomologous end-joining (NHEJ) in a ligase IV-dependent manner. SCR7 pyrazine is also a CRISPR/Cas9 enhancer which increases the efficiency of Cas9-mediated homology-directed repair (HDR). SCR7 pyrazine induces cell apoptosis and has anticancer activity[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9612
 Restricted Item