Learn More
Abcam SCR7 pyrazine, NHEJ inhibitor, 25MG
Catalog No. 502672753
Encompass_Preferred
MW 332.4 Da, Purity >98%. Small molecule nonhomologous end-joining (NHEJ) inhibitor. SCR7 analog. Enhances CRISPR-Cas9-mediated homology-directed repair (HDR) efficiency in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-2753 Supplier Abcam Supplier No. AB22237525MG
MW 332.4 Da, Purity >98%. Small molecule nonhomologous end-joining (NHEJ) inhibitor. SCR7 analog. Enhances CRISPR-Cas9-mediated homology-directed repair (HDR) efficiency in vitro.
The product is subject to the following: Abcam Restricted Use Statement