Learn More
STEMCELL Technologies SCR7, Size: 5 mg
SDP
Catalog No. 501976181
Encompass
SCR7 is an inhibitor of DNA ligase IV, which is responsible for the repair of DNA double-strandbreaks via the non-homologous end joining (NHEJ) repair pathway (Srivastava et al.). Due to its reported success in impeding cancer cell growth and potential impact on future cancer therapeutics, SCR7 has been closely studied in many recent publications (Hosoya & Miyagawa; John et al.). GENOME EDITING·Inhibits NHEJ-dependent DNA repair; this inhibition is reported to enhance precise homology-directed repair (HDR)-dependent CRISPR-Cas9 genome editing (Chu et al.; Maruyama et al.; Pinder et al.). However, these effects are cell type-specific and context-dependent (Song et al.; Xie et al.; Yang et al.; Zhang et al.).CANCER RESEARCH·Activates apoptosis of cancer cells by inhibiting DNA ligase IV to increase the efficacy of DNA double-strand break-inducing therapy (chemo- or radio-therapy) (Srivastava et al.).
Catalog No. 50-197-6181 Supplier STEMCELL Technologies Supplier No. 74102
SCR7 is an inhibitor of DNA ligase IV, which is responsible for the repair of DNA double-strandbreaks via the non-homologous end joining (NHEJ) repair pathway (Srivastava et al.). Due to its reported success in impeding cancer cell growth and potential impact on future cancer therapeutics, SCR7 has been closely studied in many recent publications (Hosoya & Miyagawa; John et al.). GENOME EDITING·Inhibits NHEJ-dependent DNA repair; this inhibition is reported to enhance precise homology-directed repair (HDR)-dependent CRISPR-Cas9 genome editing (Chu et al.; Maruyama et al.; Pinder et al.). However, these effects are cell type-specific and context-dependent (Song et al.; Xie et al.; Yang et al.; Zhang et al.).CANCER RESEARCH·Activates apoptosis of cancer cells by inhibiting DNA ligase IV to increase the efficacy of DNA double-strand break-inducing therapy (chemo- or radio-therapy) (Srivastava et al.).