Learn More
BIOHIPPO Scramble-sh GFP Blasticidin
SDP
Catalog No. 5000330798
Encompass
Scr-sh-GFP-BSD shRNA lentiviral vector expressing GFP with Blasticidin selection for studies in Each vial provides a nominal titer of 2x10E6 transducing units TU Scrambled shRNA Control Lentivirus High-titer shRNA lentiviral particles generated from a mixture of two independent scrambled shRNA control constructs The scrambled shRNA lentivirus is ultra-purified and concentrated to high-titer by PEG precipitation and sucrose gradient centrifugation and ideal for transducing difficult-to-transfect cells including thawed and/or primary cells Recommended for use as a control with any of our shRNA or miRNA lentiviral particles
Catalog No. 50-003-30798 Supplier BIOHIPPO Supplier No. BHV194001805
Scr-sh-GFP-BSD shRNA lentiviral vector expressing GFP with Blasticidin selection for studies in Each vial provides a nominal titer of 2x10E6 transducing units TU Scrambled shRNA Control Lentivirus High-titer shRNA lentiviral particles generated from a mixture of two independent scrambled shRNA control constructs The scrambled shRNA lentivirus is ultra-purified and concentrated to high-titer by PEG precipitation and sucrose gradient centrifugation and ideal for transducing difficult-to-transfect cells including thawed and/or primary cells Recommended for use as a control with any of our shRNA or miRNA lentiviral particles