Learn More
Apexbio Technology LLC Scrambled 10Panx, 5mg. Cas: 1315378-72-3 MFCD: MFCD16621675. Panx-1 mimetic inhibitory peptide, blocks pannexin-1 gap junctions
SDP
Supplier:  Apexbio Technology LLC A27015
Encompass_Preferred
Scrambled 10Panx is the scrambled form of 10Panx (WRQAAFVDSY), a mimetic peptide of pannexin 1 that inhibits dye uptake by macrophages without affecting cellular membrane currents. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4761