Learn More
Cole-Parmer Argos Technologies Cell Scraper, 39 cm L x 30 mm Blade, Multi-Position, Sterile; 100/Cs
Catalog No. 501960664 Shop All Cole-Parmer Products
Encompass_Preferred
This cell scraper has a pivot blade that can horizontally or vertically suit your needs. Access the entire cell culture surface, including difficult to reach places with the swivel blade. Prevent slippage, during use, as well as cell damage by using its handle that features a ribbed-grip design. Its TPE blade won't hurt cells. Each scraper is packaged individually and sterilized by gamma irradiation
Catalog No. 50-196-0664 Supplier Cole-Parmer Supplier No. 0439654
This cell scraper has a pivot blade that can horizontally or vertically suit your needs. Access the entire cell culture surface, including difficult to reach places with the swivel blade. Prevent slippage, during use, as well as cell damage by using its handle that features a ribbed-grip design. Its TPE blade won't hurt cells. Each scraper is packaged individually and sterilized by gamma irradiation