Learn More
TWD Scientific LLC BioX 180mm Cell Scraper. Case of 100
SDP
Catalog No. 502274280 Shop All TWD Scientific LLC Products
Encompass_Preferred
BioX disposable polyethylene sterile cell scraper (handle length: 180mm, blade length: 18mm). Scrapers come individually wrapped. 100/case, price/case
This item is not returnable. View return policy
Catalog No. 50-227-4280 Supplier TWD Scientific LLC Supplier No. BX701180
BioX disposable polyethylene sterile cell scraper (handle length: 180mm, blade length: 18mm). Scrapers come individually wrapped. 100/case, price/case