Learn More
TWD Scientific LLC BioX 250mm Cell Scraper. Case of 200
SDP
Catalog No. 502274281 Shop All TWD Scientific LLC Products
Encompass_Preferred
BioX disposable polyethylene sterile cell scraper (handle length: 250mm, blade length: 18mm). Scrapers come individually wrapped. 200/case, price/case
This item is not returnable. View return policy
Catalog No. 50-227-4281 Supplier TWD Scientific LLC Supplier No. BX701250
BioX disposable polyethylene sterile cell scraper (handle length: 250mm, blade length: 18mm). Scrapers come individually wrapped. 200/case, price/case