Learn More
BURKLE INC Scraper stainless steel, AISI 316, LxW 10.63 x 3.94 in (270 x 100 mm)
Catalog No. 5000025586
Encompass_Preferred
The high-quality scraper is suitable for scraping off, smoothing, cleaning, levelling out and also for removing strongly adhesive, sticky substances or dried up media. The edge of the corrosion-resistant stainless steel scraper is ideal for blending and portioning viscous media.
The smooth, seamless and sturdy surface ensures cleaning without difficulty and meets hygienic requirements.
The scraper makes a suitable implement for use in laboratories, production and clean rooms. It is ideal for use under rugged conditions in pharmaceuticals, chemistry, cosmetics, animal feed and food & beverage.
Catalog No. 50-000-25586 Supplier BURKLE INC Supplier No. 53860035
The high-quality scraper is suitable for scraping off, smoothing, cleaning, levelling out and also for removing strongly adhesive, sticky substances or dried up media. The edge of the corrosion-resistant stainless steel scraper is ideal for blending and portioning viscous media.
The smooth, seamless and sturdy surface ensures cleaning without difficulty and meets hygienic requirements.
The scraper makes a suitable implement for use in laboratories, production and clean rooms. It is ideal for use under rugged conditions in pharmaceuticals, chemistry, cosmetics, animal feed and food & beverage.