Learn More
BURKLE INC Scraper SteriPlast, PS, sterile, 3.15 in. (80 mm)
Catalog No. 503081221
Encompass_Preferred
Sturdy and handy scraper for disposable use. The scraper is suitable for scraping, smoothing or cleaning, as well as for removing residues.
The polystyrene scraper is produced in our class 7 (10,000) cleanroom and individually packaged for single-use.
Catalog No. 50-308-1221 Supplier BURKLE INC Supplier No. 53781030
Sturdy and handy scraper for disposable use. The scraper is suitable for scraping, smoothing or cleaning, as well as for removing residues.
The polystyrene scraper is produced in our class 7 (10,000) cleanroom and individually packaged for single-use.