Learn More
Grainger Scraper Straight SS 1 1/2 W
Catalog No. 502767424
Encompass_Preferred
The Grainger 16W161 is a 120VAC, 60Hz, 16 watt T8 fluorescent light bulb that produces 1,100 lumens of light output. This 4 foot long bulb has a medium bi-pin base and fits most standard T8 fluorescent fixtures. It has a rated life of 24,000 hours and a color temperature of 4100K. The 16W161 bulb is ideal for applications requiring bright, energy efficient fluorescent lighting such as offices, schools, retail stores, and warehouses. With its high lumen output and reliable performance, this T8 bulb from Grainger provides excellent illumination while using less energy than standard incandescent bulbs.
Catalog No. 50-276-7424 Supplier Grainger Supplier No. 16W161
The Grainger 16W161 is a 120VAC, 60Hz, 16 watt T8 fluorescent light bulb that produces 1,100 lumens of light output. This 4 foot long bulb has a medium bi-pin base and fits most standard T8 fluorescent fixtures. It has a rated life of 24,000 hours and a color temperature of 4100K. The 16W161 bulb is ideal for applications requiring bright, energy efficient fluorescent lighting such as offices, schools, retail stores, and warehouses. With its high lumen output and reliable performance, this T8 bulb from Grainger provides excellent illumination while using less energy than standard incandescent bulbs.