Learn More
Grainger Scraper Straight SS 3 W
Catalog No. 502767057
Encompass_Preferred
The Hyde Scraper Blade Material Stainless Steel is a stainless steel scraper blade that measures 3 inches wide by 18 inches long and has a thickness of 0.020 inches. It features a beveled scraping edge and is made from hardened and tempered 420 stainless steel for durability and corrosion resistance. This versatile scraper blade can be used for removing adhesives, paints, mastics and other materials from various surfaces. The thin profile allows access into tight spaces. Hyde scraper blades are made in the USA.
Catalog No. 50-276-7057 Supplier Grainger Supplier No. 16W165
The Hyde Scraper Blade Material Stainless Steel is a stainless steel scraper blade that measures 3 inches wide by 18 inches long and has a thickness of 0.020 inches. It features a beveled scraping edge and is made from hardened and tempered 420 stainless steel for durability and corrosion resistance. This versatile scraper blade can be used for removing adhesives, paints, mastics and other materials from various surfaces. The thin profile allows access into tight spaces. Hyde scraper blades are made in the USA.