Learn More
MSC 3M Sanding Screen, Silicon Carbide, 150 Grit, 17 in, Round
Catalog No. 502839079
Encompass
The sanding screen is a round, brown mesh screen disc made with Silicon Carbide abrasive material and an open mesh fabric backing. It is designed for consistent cutting performance and can be used effectively in both wet and dry applications on various surfaces such as wood, urethane finish, concrete, marble, stone, drywall, plaster, spackling compounds, metal, plastic, and painted surfaces.
  • Resists clogging for longer life
  • Rinse and reuse
  • Use both sides of screen for long life
  • Reduces loading
  • Use wet or dry
  • Mineral color will not discolor the sanding surface
  • Strong fiber and uniform open construction
  • Consistent cut from start to finish
Catalog No. 50-283-9079 Supplier MSC Supplier No. 75592675
The sanding screen is a round, brown mesh screen disc made with Silicon Carbide abrasive material and an open mesh fabric backing. It is designed for consistent cutting performance and can be used effectively in both wet and dry applications on various surfaces such as wood, urethane finish, concrete, marble, stone, drywall, plaster, spackling compounds, metal, plastic, and painted surfaces.
  • Resists clogging for longer life
  • Rinse and reuse
  • Use both sides of screen for long life
  • Reduces loading
  • Use wet or dry
  • Mineral color will not discolor the sanding surface
  • Strong fiber and uniform open construction
  • Consistent cut from start to finish