Learn More
MSC 3M Sanding Screen, 60 Grit, 16 in, Silicon Carbide
Catalog No. 502839714
Encompass
The sanding screen is a mesh disc featuring strong fiber and uniform open construction, providing a consistent cut from start to finish. It is designed for industrial and occupational use.
  • Rinsable and reusable
  • Can be used on both sides for extended life
  • Reduces loading
  • Suitable for wet or dry applications
Catalog No. 50-283-9714 Supplier MSC Supplier No. 75592667
The sanding screen is a mesh disc featuring strong fiber and uniform open construction, providing a consistent cut from start to finish. It is designed for industrial and occupational use.
  • Rinsable and reusable
  • Can be used on both sides for extended life
  • Reduces loading
  • Suitable for wet or dry applications