Learn More
Grainger Screen Frm Corners 5/16 Wht Pl PK100
Catalog No. 502768584
Encompass_Preferred
The Grainger 170F41 is a 3/4 HP, 115V, single phase, open drip proof motor with a 1075 RPM base speed. It has a 48 frame size, 1.15 service factor, and features a cast iron body with standard foot mounting. The motor is thermally protected and suitable for continuous duty applications.
Catalog No. 50-276-8584 Supplier Grainger Supplier No. 170F41
The Grainger 170F41 is a 3/4 HP, 115V, single phase, open drip proof motor with a 1075 RPM base speed. It has a 48 frame size, 1.15 service factor, and features a cast iron body with standard foot mounting. The motor is thermally protected and suitable for continuous duty applications.