Learn More
Aat Bioquest Screen Quest™ Fluo-8 No Wash Calcium Assay Kit | 10 Plates
SDP
Catalog No. NC9012743
Encompass_Preferred
Calcium flux assays are preferred methods in drug discovery for screening G protein coupled receptors (GPCR). Screen Quest™ Fluo-8 NW Calcium Assay Kit provides a homogeneous fluorescence-based assay for detecting the intracellular calcium mobilization. Cells expressing a GPCR of interest that signals through calcium are pre-loaded with our proprietary Fluo-8 NW which can cross cell membrane. Fluo-8 NW is the brightest calcium indicator available for HTS screening. Once inside the cell, the lipophilic blocking groups of Fluo-8 AM are cleaved by non-specific cell esterase, resulting in a negatively charged fluorescent dye that stays inside cells, and its fluorescence is greatly enhanced upon binding to calcium. When cells stimulated with screening compounds, the receptor signals release of intracellular calcium, which greatly increase the fluorescence of Fluo-8 NW.
Catalog No. NC9012743 Supplier Aat Bioquest Supplier No. 36315
Calcium flux assays are preferred methods in drug discovery for screening G protein coupled receptors (GPCR). Screen Quest™ Fluo-8 NW Calcium Assay Kit provides a homogeneous fluorescence-based assay for detecting the intracellular calcium mobilization. Cells expressing a GPCR of interest that signals through calcium are pre-loaded with our proprietary Fluo-8 NW which can cross cell membrane. Fluo-8 NW is the brightest calcium indicator available for HTS screening. Once inside the cell, the lipophilic blocking groups of Fluo-8 AM are cleaved by non-specific cell esterase, resulting in a negatively charged fluorescent dye that stays inside cells, and its fluorescence is greatly enhanced upon binding to calcium. When cells stimulated with screening compounds, the receptor signals release of intracellular calcium, which greatly increase the fluorescence of Fluo-8 NW.