Learn More
Ace Glass, Inc. SCREEN SUPPORT SCREEN SUPPORT
SDP
Catalog No. 50975467
Encompass_Preferred
Screen support for catalyst column for the ACE-Burlitch system. The ACE-Burlitch inert atmosphere system is designed for maximum convenience and versatility in the handling of air-sensitive compounds in conjunction with ACE no-air labware, or suitiably adapted conventional glassware.
Catalog No. 50-975-467 Supplier Ace Glass, Inc. Supplier No. 781835
Screen support for catalyst column for the ACE-Burlitch system. The ACE-Burlitch inert atmosphere system is designed for maximum convenience and versatility in the handling of air-sensitive compounds in conjunction with ACE no-air labware, or suitiably adapted conventional glassware.