Learn More
FUJIFILM Biosciences ScreenFect A
Catalog No. 5000450297
Encompass_Preferred
ScreenFect A is a low-toxicity transfection reagent made of novel cationic lipids effective for DNA and siRNA delivery across various eukaryotic cell types including common cell lines stem cells blood cells microglia and primary cultures It can be directly added to media containing antibiotics and serum without needing medium change post-transfection and contains no harmful substances High transfection efficiency non-hazardous includes dilution buffer easy to use suitable for various mammalian cell lines For research use only RUO
Catalog No. 50-004-50297 Supplier FUJIFILM Biosciences Supplier No. 29973203
ScreenFect A is a low-toxicity transfection reagent made of novel cationic lipids effective for DNA and siRNA delivery across various eukaryotic cell types including common cell lines stem cells blood cells microglia and primary cultures It can be directly added to media containing antibiotics and serum without needing medium change post-transfection and contains no harmful substances High transfection efficiency non-hazardous includes dilution buffer easy to use suitable for various mammalian cell lines For research use only RUO