Learn More
Molecular Devices Molecular Devices ImageXpress Nano, Imaging System, 4-Position Objective, 5-Channel Filter
Supplier:  Molecular Devices IXNDB
Encompass_Preferred
The Molecular Devices ImageXpress Nano is a compact automated fluorescence high-content imaging system for medium-throughput microscopy assays. It combines a solid-state light source, precision motorized stage and autofocus, multi-position optics and integrated acquisition software for 2D and 3D imaging.
  • Four-position objective changer with configurable objectives.
  • Five-position fluorescence filter cube changer supporting up to five channels.
  • CMOS camera for fluorescence image capture.
  • Integrated image acquisition and analysis software.
  • Supports microscope slides and plate formats up to 1536 wells.
  • Includes accessory kit with calibration slides and test plates.
  • Optional environmental control for temperature, humidity, and CO2.
Catalog No. 50-000-14669
Only null left
Add to Cart
Request a Quote