Learn More
Molecular Devices Molecular Devices ImageXpress Micro Confocal, Spinning Disk, Widefield, High Throughput
Supplier:  Molecular Devices IXMCDB
Encompass_Preferred
The Molecular Devices ImageXpress Micro Confocal is a high-content imaging system that combines spinning-disk confocal and widefield modalities for automated acquisition and analysis of live and fixed cell assays. It supports high-throughput multiwell plate imaging, z-stack 3D imaging, and integration with automated workflows for screening applications.
  • Spinning-disk confocal and widefield fluorescence, brightfield, and phase contrast imaging.
  • sCMOS-based detection for high sensitivity and fast acquisition.
  • Supports multiwell plates up to 1536-well and microscope slides.
  • Z-stack acquisition and 3D image analysis for spheroids and thick tissues.
  • Environmental control options for temperature and gas regulation during live-cell imaging.
  • Integrated acquisition and analysis software with AI-driven analysis options.
  • Optional onboard fluidics for compound addition and media exchange.
Catalog No. 50-000-14701
Add to Cart
Request a Quote