Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One SCREENSTAR 1536-well, Cell Culture-Treated, Flat-Bottom Microplate
GREENER_CHOICE

Catalog No. 07000930 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Black, Clear Bottom
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07-000-930 Black, Clear Bottom
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-000-930 Supplier Greiner Bio-One Supplier No. 789866
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Color:Black, Clear Bottom; Sterility:Sterile; Lid:Without Lid

  • Recessed film bottom facilitates use of lenses with low working distance, high aperture
  • Cycloolefin film bottom with 190μm thickness
  • Superior film quality enables high-quality images at high magnification
  • Protective film
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Lid Without Lid
Culture Area 1 to 2 mm2
Array 32 x 48
Autoclavable Not Autoclavable
Packaging 17/Pk., 68/Cs.
Color Black, Clear Bottom
Material Cycloolefin
No. of Wells 1536
Sterility Sterile
Surface Treatment Cell Culture
Total Volume (Metric) Well 8 to 17 μL
Well Shape Rounded Square
Dnase Free No
Max. RCF 1500 x g (swinging-bucket rotor)
RNase Free No
Temperature Range (Metric) -20°C to +60°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.