Learn More
Tisch Scientific Screw Cap 13mm Black Plastic Vial Cap with Red PTFE/White Silicone Septa - 100/pk, CV1945
SDP
Catalog No. NC2064329
Encompass
Black polypropylene screw cap, 8.5mm center hole, 13mm diameter, with red PTFE/white silicone septa, 100/pk, CV1945
Quantity per Pack100
Cap TypeScrew Cap
Cap Size13mm
ColorBlack
MaterialPolypropylene
Pre-slitNo
Center Hole Size8.5mm
Septa TypeRed PTFE/White Silicone
Catalog No. NC2064329 Supplier Tisch Scientific Supplier No. CV1945
Black polypropylene screw cap, 8.5mm center hole, 13mm diameter, with red PTFE/white silicone septa, 100/pk, CV1945
Quantity per Pack100
Cap TypeScrew Cap
Cap Size13mm
ColorBlack
MaterialPolypropylene
Pre-slitNo
Center Hole Size8.5mm
Septa TypeRed PTFE/White Silicone