Learn More
Alkali Scientific ASI™ Screw-Cap 5mL MacroTubes, 5mL Screw-cap MacroTube Vivid™, sterile, w/ vivid printed graduation w/ attached screw caps in foam racks, 500/Case
SDP
Catalog No. 503026459
Encompass_Preferred
ASI Screw-Cap 5mL MacroTubes Alkali Scientific’s MacroTubeª 5mL Centrifuge Tubes are compatible with standard centrifuges and have exceptional clarity. Useful from -86°C to 110°C, these conical-bottom tubes are excellent for boiling and for sample preservation at very low temperatures. They come as gamma sterilized in racked packaging with attached caps or in non-sterile bags with separate caps. Our foam racks are similar to the packaging normally associated with 15mL tubes (upright with caps attached), but they require about 60% less storage space. Gamma sterilized, racked conical tubes or non-sterile bags Exceptional clarity RNase, DNase & Pyrogen FREE Compatible with standard centrifuges Withstands up to 25,000xG Dual graduations at every 0.2mL and 0.5mL
Catalog No. 50-302-6459 Supplier Alkali Scientific Supplier No. C2545
ASI Screw-Cap 5mL MacroTubes Alkali Scientific’s MacroTubeª 5mL Centrifuge Tubes are compatible with standard centrifuges and have exceptional clarity. Useful from -86°C to 110°C, these conical-bottom tubes are excellent for boiling and for sample preservation at very low temperatures. They come as gamma sterilized in racked packaging with attached caps or in non-sterile bags with separate caps. Our foam racks are similar to the packaging normally associated with 15mL tubes (upright with caps attached), but they require about 60% less storage space. Gamma sterilized, racked conical tubes or non-sterile bags Exceptional clarity RNase, DNase & Pyrogen FREE Compatible with standard centrifuges Withstands up to 25,000xG Dual graduations at every 0.2mL and 0.5mL