Learn More
Bellco Glass, Inc. Screw Cap Assembly ,38mm
SDP
Catalog No. 501945149
Encompass_Preferred
Black Phenolic Screw Cap Assembly For Adjustable Hanging Bar Spinner FlaskBlack phenolic screw caps are manufactured from specially formulated phenolic resin to resist the effects of temperature and steam autoclaving. They have a non-toxic white rubber liner for all general culture applications. Consists of screw cap liner and stopper or compression fitting. 38mm.
Catalog No. 50-194-5149 Supplier Bellco Glass, Inc. Supplier No. 196790038
Black Phenolic Screw Cap Assembly For Adjustable Hanging Bar Spinner FlaskBlack phenolic screw caps are manufactured from specially formulated phenolic resin to resist the effects of temperature and steam autoclaving. They have a non-toxic white rubber liner for all general culture applications. Consists of screw cap liner and stopper or compression fitting. 38mm.