Learn More
MilliporeSigma™ Supelco™ Screw Cap Black Polypropylene
Catalog No. 111011218 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
4 mL Vial, 13-425 Thread
Material:
PTFE/Silicone, Black Polypropylene (Top Hat)
Catalog No. For Use With (Equipment) Material
11-101-1218 4 mL Vial, 13-425 Thread PTFE/Silicone, Black Polypropylene (Top Hat)
1 options
Catalog No. 11-101-1218 Supplier MilliporeSigma™ Supelco™ Supplier No. 27439
Add to Cart
Add to Cart
Screw caps are available as hole caps for use with sealing septa.
  • Used for capping water samples in amber glass vial comprising of a polytetrafluoroethylene silicon septum for the analysis of chlorinated volatile organic compounds in water samples using headspace-solid-phase microextraction with gas chromatography (HS-SPME-GC).
  • Screw caps (large, opening in top of cap, no septum, thread 8-425) are available as hole caps for use with sealing septa.
  • Polypropylene caps can withstand a maximum temperature of 135°C.
TRUSTED_SUSTAINABILITY
Specifications
Material PTFE/Silicone, Black Polypropylene (Top Hat)
No. per Pack 100
For Use With (Equipment) 4 mL Vial, 13-425 Thread