Learn More
Agilent Technologies SCREW CAP, BONDED, BLUE, PTFE/
Catalog No. NC1910960
Encompass
Screw cap, bonded, blue, PTFE/white silicone septa, 100/pk. Cap size: 12 mm
Catalog No. NC1910960 Supplier Agilent Technologies Supplier No. 51907021
Screw cap, bonded, blue, PTFE/white silicone septa, 100/pk. Cap size: 12 mm