Learn More
PROANALYTICS LLC SCREW CAP -BUTYL/PTFE
SDP
Catalog No. NC2235254
Encompass
Securecell:Numera:Consumables:ND18 Screw Cap -Butyl/PTFE 941781 ND18 Magnetic Screw Cap, 8mm Hole, Butyl/PTFE Septa, 100 Pack 23.01
This item is not returnable. View return policy
Catalog No. NC2235254 Supplier PROANALYTICS LLC Supplier No. 941781
Securecell:Numera:Consumables:ND18 Screw Cap -Butyl/PTFE 941781 ND18 Magnetic Screw Cap, 8mm Hole, Butyl/PTFE Septa, 100 Pack 23.01