Learn More
MTC Bio Culture Tube, 14mL, 17 x 100mm, PS, w/ attached 2-position screw-cap, printed graduations, sterile, 20 bags of 25 tubes, 500/case
Catalog No. 502268504
Encompass_Preferred
MTC Bio’s new DuoClick™ tubes feature a unique, patent pending screw closure system. DuoClick tubes completely eliminate the excess force, splashing and general frustration of traditional snap-cap culture tubes. The advanced cap design includes two distinct positions for aerobic and anaerobic culturing. Just rotate the cap 1/4 turn to the first click to secure the cap in the aerobic position. Or, rotate it another 1/2 turn past the second click for a leakproof, airtight seal for anaerobic culturing. DuoClick tubes are available gamma sterilized, with screw caps attached, and in our exclusive ready-to-use rack packaging. Tubes feature clear printed graduations, and are sterilized by electron beam. RNase, DNase, pyrogen, & endotoxin free.
This item is not returnable. View return policy
Catalog No. 50-226-8504 Supplier MTC Bio Supplier No. T8840
MTC Bio’s new DuoClick™ tubes feature a unique, patent pending screw closure system. DuoClick tubes completely eliminate the excess force, splashing and general frustration of traditional snap-cap culture tubes. The advanced cap design includes two distinct positions for aerobic and anaerobic culturing. Just rotate the cap 1/4 turn to the first click to secure the cap in the aerobic position. Or, rotate it another 1/2 turn past the second click for a leakproof, airtight seal for anaerobic culturing. DuoClick tubes are available gamma sterilized, with screw caps attached, and in our exclusive ready-to-use rack packaging. Tubes feature clear printed graduations, and are sterilized by electron beam. RNase, DNase, pyrogen, & endotoxin free.