Learn More
SAI Infusion Technologies 1.3ml K3 EDTA Blood Tubes with Screw Cap
SDP
Catalog No. NC1049697
Encompass
1.3ml K3EDTA Blood Tube with Screw Cap. Ideal for collection of up to 1.3ml of blood for plasma. Slim Tube and Cap profiles ensure a perfect fit in standard racks and centrifuges. 100/pk. Product is for laboratory animal research only.
Catalog No. NC1049697 Supplier SAI Infusion Technologies Supplier No. PMTSE1.3
1.3ml K3EDTA Blood Tube with Screw Cap. Ideal for collection of up to 1.3ml of blood for plasma. Slim Tube and Cap profiles ensure a perfect fit in standard racks and centrifuges. 100/pk. Product is for laboratory animal research only.