Learn More
Neta Scientific Agilent Screw cap red 100/PK
SDP
Catalog No. 50929409
Encompass_Preferred
This item is manufactured by Agilent
Agilent Cap, screw, red, PTFE/red silicone, 100/pk. Cap size: 12 mm - 2 ml Screw Top Vials & Caps
UNSPSC Code  41121800
- 58
Catalog No. 50-929-409 Supplier Neta Scientific Supplier No. 51820719
This item is manufactured by Agilent
Agilent Cap, screw, red, PTFE/red silicone, 100/pk. Cap size: 12 mm - 2 ml Screw Top Vials & Caps
UNSPSC Code  41121800
- 58