Learn More
Chemglass Life Sciences Screw Cap, Open Top, 24-400, Green, with PTFE Faced Silicone Septa
SDP
Catalog No. CG491004
Encompass_Preferred
Screw thread size: 24-400, GREEN. Open top polypropylene screw caps supplied with PTFE faced silicone septa. Supplied in packages of 144.
Catalog No. CG491004 Supplier Chemglass Life Sciences Supplier No. CG491004
Screw thread size: 24-400, GREEN. Open top polypropylene screw caps supplied with PTFE faced silicone septa. Supplied in packages of 144.