Learn More
Sarstedt Inc SCREW CAP TUBE, 8 ML,1000/CS
Supplier:  Sarstedt Inc 62.542.009
Encompass_Preferred
Screw cap tube, working volume: 8 ml, (LxØ): 57 x 16.5 mm. material: PP, flat base, transparent, screw cap, HD-PE, cap: natural, cap assembled, with print, label / print: white, with graduations, 1000 piece(s)/bag
Catalog No. NC2066052