Learn More
USA Scientific Inc 2.0 mL self-standing screw cap tube, mixed color caps
Catalog No. NC9489374
Encompass_Preferred
2.0 mL self-standing screw cap microcentrifuge tube, graduated, natural with mixed color caps, 500/pack
Catalog No. NC9489374 Supplier USA Scientific Inc Supplier No. 14209799
2.0 mL self-standing screw cap microcentrifuge tube, graduated, natural with mixed color caps, 500/pack