Learn More
Neta Scientific Agilent Vial,screw,2ml,clr,cert,100PK
SDP
Catalog No. 50929167
Encompass_Preferred
This item is manufactured by Agilent
Agilent Vial, screw top, clear, certified, 2 mL, 100/pk. Vial size: 12 x 32 mm (12 mm cap) - 2 ml Screw Top Vials & Caps
UNSPSC Code  41121806
- 58
Catalog No. 50-929-167 Supplier Neta Scientific Supplier No. 51820714
This item is manufactured by Agilent
Agilent Vial, screw top, clear, certified, 2 mL, 100/pk. Vial size: 12 x 32 mm (12 mm cap) - 2 ml Screw Top Vials & Caps
UNSPSC Code  41121806
- 58