Learn More
Metrohm USA SCREW CAP W/HOLE FOR SEPTUM
SDP
Supplier:  Metrohm USA 62701040
Encompass
Made of PBT. With a hole for the 6.1448.020 septum. For the 6.5405.000 cell without diaphragm and the 6.1464.32X and 6.1465.320 titration vessels.
Catalog No. NC0084776