Learn More
Bellco Glass, Inc. Screw Cap w/Rubber Liner, 38mm Short Skirted
SDP
Catalog No. 501946064
Encompass_Preferred
Autoclavable black mineral filled phenolic resin with white rubber liner. The screw cap is short skirted for use with glass roller bottles.. MADE IN THE USA Thread: 38-400 Case quantity: 36
Catalog No. 50-194-6064 Supplier Bellco Glass, Inc. Supplier No. 773000038
Autoclavable black mineral filled phenolic resin with white rubber liner. The screw cap is short skirted for use with glass roller bottles.. MADE IN THE USA Thread: 38-400 Case quantity: 36