Learn More
Bellco Glass, Inc. Screw Cap w/Rubber Liner, 51mm Short Skirted
SDP
Catalog No. 501945332
Encompass_Preferred
Manufactured from black mineral-filled phenolic resign with white rubber liner. Short skirt for use with gas roller bottles. Autoclavable, black mineral-filled phenolic resin with white rubber liner Short skirt for use with glass roller bottles. THREAD:51-400. CASE QTY:72.
Catalog No. 50-194-5332 Supplier Bellco Glass, Inc. Supplier No. 202900051
Manufactured from black mineral-filled phenolic resign with white rubber liner. Short skirt for use with gas roller bottles. Autoclavable, black mineral-filled phenolic resin with white rubber liner Short skirt for use with glass roller bottles. THREAD:51-400. CASE QTY:72.