Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One Screw Caps for Polystyrene Roller Bottles
GREENER_CHOICE

Catalog No. 07000544 Shop All Greiner Bio-One Products
Change view
Click to view available options
Thread Size (Metric):
83 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size (Metric)
07-000-544 83 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-000-544 Supplier Greiner Bio-One Supplier No. 383361
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For polystyrene roller bottle. Greiner Bio-One Screw Caps for Polystyrene Roller Bottles are sterile and is free of detectable DNase, RNase, human DNA.

  • Sterile
  • Non-cytotoxic
  • Non-pyrogenic
  • Free of detectable DNase, RNase, human DNA
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Material HDPE
No. per Case 100
Sterility Sterile
Thread Size (Metric) 83 mm
Neck Finish 83 mm
For Use With (Equipment) Polystyrene roller bottle
Autoclavable Not Autoclavable
Disposable Single-use
Product Type Screw Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.