Learn More
Krackeler Scientific Screw Caps with Septa
SDP
Catalog No. 51820717N
Change view
Click to view available options
Color:
Blue
Blue, Red
Blue, White
Septum:
PTFE/Rubber
PTFE/Silicone
PTFE/Silicone, Bonded
PTFE/Silicone/PTFE
Pre-slit:
No
Yes
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Septum Pre-slit
51820717N Blue, Red PTFE/Rubber No
056005200N Blue PTFE/Silicone/PTFE No
056005202N Blue PTFE/Silicone Yes
05601697N Blue PTFE/Silicone, Bonded Yes
HP51820720N Blue, White PTFE/Silicone No
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 51820717N Supplier Krackeler Scientific Supplier No. 9251820717EA
Add to Cart
Add to Cart
For wide opening screw top vials
Specifications
Material (Liner) PTFE/Rubber
Color Blue, Red
Material Polypropylene
Pre-slit No
Septum PTFE/Rubber