Learn More
Krackeler Scientific Screw Caps with Septa
Description
- Supplied with a PTFE-faced septum.
- Compatible with wide-opening chromatography vials.
Specifications
Specifications
| Material (Liner) | PTFE/Silicone |
| Color | Blue, White |
| Material | Polypropylene |
| Pre-slit | No |
| Septum | PTFE/Silicone |
| Quantity | 100 Pk. |