Learn More
Ops Diagnostics SCREW CAPS UNLINED 240/PK
Catalog No. NC0827170
Encompass
Unlined polypropylene 15-425 screw caps for 4 ml vials. Bag of 240.
This item is not returnable. View return policy
Catalog No. NC0827170 Supplier Ops Diagnostics Supplier No. CPUL 0424007
Unlined polypropylene 15-425 screw caps for 4 ml vials. Bag of 240.