Learn More
Electron Microscopy Sciences Screw Cap Centrifuge Tube 15 ml, Polypropylene, Sterile
SDP
Supplier:  Electron Microscopy Sciences 6476401
Encompass_Preferred
Premium quality general purpose centrifuge tubes with printed graduations. Polypropylene has excellent chemical resistance and can withstand 6400 RCF.
Material: Polypropylene
Sterile: Sterile
Catalog No. 50-243-62