Learn More
Electron Microscopy Sciences Screw Cap Centrifuge Tube 15 ml, Polystyrene, Sterile
SDP
Catalog No. 5024358
Encompass_Preferred
Premium quality general purpose centrifuge tubes with printed graduations. Polystyrene offers glass-like clarity and can withstand 1800RCF.
Material: Polystyrene
Sterile: Sterile
Catalog No. 50-243-58 Supplier Electron Microscopy Sciences Supplier No. 6476301
Premium quality general purpose centrifuge tubes with printed graduations. Polystyrene offers glass-like clarity and can withstand 1800RCF.
Material: Polystyrene
Sterile: Sterile