Learn More
AVANTOR MASTERFLEX (VWR LLC) Bel-Art Flow Control Clamp, Delrin Screw Clamp, 3/Pk
Catalog No. NC9560174
Encompass
DELRIN(R) resists weak acids, bases, and organic solvents. Controls flow in flexible tubing up to 1/2" (13 mm) OD. Replaces stopcocks in many applications. Install without disconnecting tubing. Autoclavable. Bel-Art.
This item is not returnable. View return policy
Catalog No. NC9560174 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0683310
DELRIN(R) resists weak acids, bases, and organic solvents. Controls flow in flexible tubing up to 1/2" (13 mm) OD. Replaces stopcocks in many applications. Install without disconnecting tubing. Autoclavable. Bel-Art.