Learn More
Electron Microscopy Sciences Tripod Polisher Screw Clamp "L" Bracket
SDP
Catalog No. 50980868
Encompass
Used with Pyrex Insert for Wedge Polishing (#50116-60), SEM Stub (#50116-65) or Wedge Polishing Mount (#50116-61). Can be used for polishing edge of sample to area for SEM or preparation for final wedge polishing for TEM.
Catalog No. 50-980-868 Supplier Electron Microscopy Sciences Supplier No. 5011625
Used with Pyrex Insert for Wedge Polishing (#50116-60), SEM Stub (#50116-65) or Wedge Polishing Mount (#50116-61). Can be used for polishing edge of sample to area for SEM or preparation for final wedge polishing for TEM.